Order Entry
Germany
ContactUsLinkComponent
4-Amino-2,3,5,6-tetrafluoropyridine 98%
Toxic
Irritant
4-Amino-2,3,5,6-tetrafluoropyridine 98%
Catalog # B23815.06
CAS Number:  
Toxic
Irritant
4-Amino-2,3,5,6-tetrafluoropyridine 98%
Catalog # B23815.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :166.08 g/mol
Melting Pt:84…87 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:1682-20-8
Formula:C₅H₂F₄N₂
MDL Number:MFCD00010855