Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Heptyltrichlorosilane
Heptyltrichlorosilane
Heptyltrichlorosilane
Heptyltrichlorosilane
Catalog # SILNSILS09305.5
Supplier:
CHEMPUR SILANE
CAS Number:
Expand image
Heptyltrichlorosilane
Catalog # SILNSILS09305.5
Supplier:
CHEMPUR SILANE
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
Specifications
About this item
Product Details
:
EINECS:
212-807-7
CAS Number:
871-41-0
Formula:
C₇H₁₅Cl₃Si
Product Details