4'-Ethoxy-2'-hydroxyacetophenone, Sigma-Aldrich®
Catalog # SIAL579173-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:180.200 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:37470-42-1
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00100624