Buffers, MOPS, 1,0 M solutions
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Specifications
- CAS No.:1132-61-2
- MDL No.:MFCD00006183
- Physical form:Liquid
- Odour/scent:Odourless
- Appearance:Colourless
- Storage temperature:2°C to 8°C