Order Entry
Germany
ContactUsLinkComponent
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde for synthesis
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde for synthesis
Catalog # 8.14808.0010
Supplier:  Merck
CAS Number:  
5-(4-Fluorophenyl)-3-pyridinecarbaldehyde for synthesis
Catalog # 8.14808.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :201.2000 g/mol
CAS Number:381684-96-4
Formula:C₁₂H₈FNO
MDL Number:MFCD02093924