Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Trifluormethyl)nicotinonitrile
6-(Trifluormethyl)nicotinonitrile
6-(Trifluormethyl)nicotinonitrile
6-(Trifluormethyl)nicotinonitrile
Supplier:
FLUOROCHEM
Certificates
Expand image
6-(Trifluormethyl)nicotinonitrile
Supplier:
FLUOROCHEM
About this item
CAS Number:
216431-85-5
:
Formula:
C₇H₃F₃N₂
MDL Number:
MFCD03001347
:
172.1100 g/mol
UN:
3439
ADR:
6.1,III