Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
Supplier:
BIONET RESEARCH
Certificates
Expand image
2-Morpholino-3-pyridinamine
Supplier:
BIONET RESEARCH
About this item
CAS Number:
51627-47-5
:
Formula:
C₉H₁₃N₃O
MDL Number:
MFCD03001243