2'-(3-Fluorobenzyloxy)acetophenone
Catalog # FLUO023221-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
CAS Number:845823-26-9
Formula:C₁₅H₁₃FO₂
MDL Number:MFCD06411645