4'-Phenoxy-2,2,2-trifluoroacetophenone
Catalog # FLUO023143-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:266.215 g/mol
CAS Number:70783-32-3
Formula:C₁₄H₉F₃O₂
MDL Number:MFCD00042392