Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
10,12-Docosadiynedioic acid
10,12-Docosadiynedioic acid
10,12-Docosadiynedioic acid
10,12-Docosadiynedioic acid
Supplier:
FLUOROCHEM
Certificates
Expand image
10,12-Docosadiynedioic acid
Supplier:
FLUOROCHEM
About this item
CAS Number:
28393-02-4
:
Formula:
C₂₂H₃₄O₄
MDL Number:
MFCD00078306
:
362.51 g/mol