Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
10,12-Docosadiynedioic acid
10,12-Docosadiynedioic acid 95%
10,12-Docosadiynedioic acid 95%
10,12-Docosadiynedioic acid 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
10,12-Docosadiynedioic acid 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
28393-02-4
:
Formula:
C₂₂H₃₄O₄
MDL Number:
MFCD00078306
:
362.51 g/mol
Melting Pt:
110…112 °C
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAL10647
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation