Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Boc-amino)piperidine
4-(Boc-amino)piperidine, Sigma-Aldrich®
4-(Boc-amino)piperidine, Sigma-Aldrich®
4-(Boc-amino)piperidine, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
4-(Boc-amino)piperidine, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
73874-95-0
:
Formula:
C₁₀H₂₀N₂O₂
MDL Number:
MFCD00798171
:
M-Clarity™ quality level = MQ100
:
200.28 g/mol