Order Entry
Germany
Orders LinkContactUsLinkComponent
Forskolin

Forskolin

Glass bottle for solids, 5 mg
Catalog # UPST19-140
Marketsource
Supplier:   UPSTATE TECHNOLOGY
Forskolin
Catalog # UPST19-140
Supplier:  UPSTATE TECHNOLOGY
Marketsource
CAS Number:  66575-29-9
Forskolin

Glass bottle for solids, 5 mg

CAS Number:  66575-29-9
Price
Inquire For Price
Log in for availability
1 * 5 mg
Product Details & Documents
Synonyms:  (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
  • CAS Number:66575-29-9
  • Formula:C₂₂H₃₄O₇
  • MDL Number:MFCD00082317
  • Molecular Weight:410.51 g/mol
  • EINECS:266-410-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.