Order Entry
Germany
ContactUsLinkComponent
2-Sulphobenzoic cyclic anhydride for synthesis, Sigma-Aldrich®
Irritant
2-Sulphobenzoic cyclic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41501.0025
Supplier:  Merck
CAS Number:  
Irritant
2-Sulphobenzoic cyclic anhydride for synthesis, Sigma-Aldrich®
Catalog # 8.41501.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    116 - 122 °C
  • Molar Mass:
    184.17 g/mol

Specification Test Results

About this item

 :
 :184.17 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ200
EINECS:201-322-6
UN:3261
 :GHS07
Storage Temperature:Ambient
CAS Number:81-08-3
Formula:C₇H₄O₄S
MDL Number:MFCD00005879