Order Entry
Germany
ContactUsLinkComponent
Lansoprazole
Lansoprazole
Catalog # EHERC14592500
Supplier:  EHRENSTORFER
CAS Number:  
Lansoprazole
Catalog # EHERC14592500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :369.36 g/mol
CAS Number:103577-45-3
Formula:C₁₆H₁₄F₃N₃O₂S
MDL Number:MFCD00866873