Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(-)-Luciferin
D-(-)-Luciferin, Sigma-Aldrich®
D-(-)-Luciferin, Sigma-Aldrich®
D-(-)-Luciferin, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
D-(-)-Luciferin, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
2591-17-5
:
Formula:
C₁₁H₈N₂O₃S₂
MDL Number:
MFCD00042929
:
280.3228 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
219-981-3