D(-)-Luciferin
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
About this item
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- EINECS:219-981-3
3 Options Available Below
Product Details & Documents
Synonyms: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
- CAS Number:2591-17-5
- Formula:C₁₁H₈N₂O₃S₂
- MDL Number:MFCD00042929
- Molecular Weight:280.3228 g/mol
- EINECS:219-981-3
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



