Order Entry
Germany
Orders LinkContactUsLinkComponent
D(-)-Luciferin

D(-)-Luciferin

Irritant
D(-)-Luciferin
Certificates
D(-)-Luciferin

2 Options Available Below

Product Details & Documents
Synonyms:  D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
  • CAS Number:2591-17-5
  • Formula:C₁₁H₈N₂O₃S₂
  • MDL Number:MFCD00042929
  • Molecular Weight:280.3228 g/mol
  • Melting Pt:200 °C
  • Storage Temperature:Freezer
  • EINECS:219-981-3

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤2.0 % (K.F.)
  • HPLC:
    ≥97.5 %
  • UV Lambda max:
    327 to 331 nm (c=1, CH3OH)
  • Specific optical rotation:
    -32.0° to -36.0° (20°C, 589 nm) (c=1, DMF)
  • Molar extinction coefficient:
    ≥18200 l/(mol.cm) (327 to 331 nm, c=1, CH3OH)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 193,00 €
    193,00 €
    1 * 10 mg
       193,00 €

  • Glass bottle
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 663,00 €
    663,00 €
    1 * 50 mg
       663,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.