Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Folic acid
Folic acid
Folic acid
Folic acid
Catalog # RBIOP1001
Supplier:
R BIOPHARM
CAS Number:
Expand image
Folic acid
Catalog # RBIOP1001
Supplier:
R BIOPHARM
CAS Number:
Specifications
Pk:
1 SET
Seq:
0125
Specifications
About this item
Product Details
:
:
441.41 g/mol
EINECS:
200-419-0
CAS Number:
59-30-3
Formula:
C₁₉H₁₉N₇O₆
MDL Number:
MFCD00079305
Product Details