Folic acid 97.0-102.0% (by HPLC) for biochemistry, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:59-30-3
:
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305
:441.41 g/mol
Melting Pt:250 °C
Density:110 kg/m³
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-419-0
Specification Test Results
- Assay (HPLC, Calc.on anhydrous substance):97.0 - 102.0%
- Identity (HPLC):Conforms
- Identity (IR-spectrum):Conforms
- Absorption ratio (A₂₅₆/A₃₆₅):2.80 - 3.00
- Spec. rotation (α 20/D; 10 g/l; NaOH 0,1 mol/l; calc. on anhydrous subst.):+18 - +22°
- Related substances (HPLC) (N-(4-aminobenzoyl)-L-glutamic acid (impurity A)):≤0.5%
- Related substances (HPLC) (Pteroic Acid (Impurity D)):≤0.6%
- Related Compounds (HPLC) (Total unspecified impurities):≤1.0%
- Water (According to Karl Fischer):5.0 - 8.5%