Folic acid
Catalog # FITZ10-F19-M608297
Supplier: Fitzgerald Industries
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:441.41 g/mol
EINECS:200-419-0
CAS Number:59-30-3
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305