Folic acid 97%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:59-30-3
:
Formula:C₁₉H₁₉N₇O₆
MDL Number:MFCD00079305
:441.41 g/mol
Melting Pt:250 °C (dec.)
Storage Temperature:Ambient
EINECS:200-419-0
Specification Test Results
- Appearance:Yellow to orange Crystalline powder
- Infrared spectrum:Conforms
- Water:≤8.5 %
- Sulfated ash:≤0.2 %
- HPLC:≥96.0 %