Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Carbamazepine 10 µg/ml in methanol
Organic reference standard, Carbamazepine 10 µg/ml in methanol
  PROONE9228
 :  
Organic reference standard, Carbamazepine 10 µg/ml in methanol
  PROONE9228
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    298-46-4
  • Description:
    Carbamazepine 10 µg/ml in methanol
  • Formula:
    C₁₅H₁₂N₂O
  • Description:
    Carbamazepine