Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer, TRIS solution pH 8,8 (1 mol/L)

TRIS buffer solution pH 8,8 (1 mol/l), Glass bottle, 1 L
Catalog # SERA39794.01
Marketsource
Supplier:   Serva
Buffer, TRIS solution pH 8,8 (1 mol/L)
Catalog # SERA39794.01
Supplier:  Serva
Marketsource
CAS Number:  
Buffer, TRIS solution pH 8,8 (1 mol/L)

TRIS buffer solution pH 8,8 (1 mol/l), Glass bottle, 1 L

CAS Number:  
Price
103,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TRIS buffer solution pH 8,8 (1 mol/l)

Specifications