Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Cloning Buffers
Buffer, TRIS solution pH 8,0 (1 mol/L), Sigma-Aldrich®
Buffer, TRIS solution pH 8,0 (1 mol/L), Sigma-Aldrich®
Buffer, TRIS solution pH 8,0 (1 mol/L), Sigma-Aldrich®
Catalog # SIALT3038-1L
Supplier:
Merck
CAS Number:
Certificates
Expand image
Buffer, TRIS solution pH 8,0 (1 mol/L), Sigma-Aldrich®
Catalog # SIALT3038-1L
Supplier:
Merck
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle
Pk:
1 L
Description:
TRIS buffer solution pH 8,0 (1 mol/l)
M-Clarity quality level:
MQ200
Specifications
About this item
Product Details
Documentation
Product Details
Documentation