Order Entry
Germany
ContactUsLinkComponent
Buffers, sodium phosphate, 0,5 M solutions
Buffers, sodium phosphate, 0,5 M solutions
Certificates
Buffers, sodium phosphate, 0,5 M solutions
Certificates

Specifications

  • MDL No.:
    MFCD00146206
  • Physical form:
    Liquid

Specifications