Order Entry
Germany
ContactUsLinkComponent
Buffer, sodium acetate, 3M aq, soln,, pH 4,5, autoclaved
Buffer, sodium acetate, 3M aq, soln,, pH 4,5, autoclaved
Certificates
Buffer, sodium acetate, 3M aq, soln,, pH 4,5, autoclaved
Certificates

Specifications

  • CAS No.:
    127-09-3
  • MDL No.:
    MFCD00012459
  • Formula weight:
    82.03
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Molecular formula:
    C2H3NaO2

Specifications

Related Information

Frequently Bought Together