Buffers, AMPSO, 0,2 M solutions
CAS No.: 102029-60-7
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00067500
- CAS No.:102029-60-7
- MDL No.:MFCD00067500
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Odour/scent:Odourless
- Appearance:Colourless