3-(1,3-Dioxolan-2-yl)propiophenone
Glass bottle for solids, 1 g
Catalog # FLUO022277-1G
Marketsource
Supplier: FLUOROCHEM
3-(1,3-Dioxolan-2-yl)propiophenone
Catalog # FLUO022277-1G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 54743-42-9
Jump to:
Product Details & Documents
Synonyms: 3-(1,3-Dioxolan-2-yl)-1-phenylpropan-1-one
- CAS Number:54743-42-9
- Formula:C₁₂H₁₄O₃
- MDL Number:MFCD06201162
- Molecular Weight:206.237 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



