3-(1,3-Dioxolan-2-yl)-4'-propylpropiophenone
Catalog # FLUO022300-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:842124-08-7
Formula:C₁₅H₂₀O₃
MDL Number:MFCD06201175