N-Boc-glycine (boc-gly-OH)
Catalog # MOLE46029276-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:175.2 g/mol
EINECS:224-864-5
CAS Number:4530-20-5
Formula:C₇H₁₃NO₄
MDL Number:MFCD00002690