Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chlorate
Sodium chlorate 50 g/l in aqueous solution, extra pure
Sodium chlorate 50 g/l in aqueous solution, extra pure
Sodium chlorate 50 g/l in aqueous solution, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Sodium chlorate 50 g/l in aqueous solution, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
7775-09-9
:
Formula:
ClNaO₃
MDL Number:
MFCD00003479
:
106.44 g/mol
EINECS:
231-887-4