Acetate standard solution, 1000 mg/l AcO- in Water (from NaOOCCH₃) standard for ion chromatography
Lieferant: Bernd Kraft
Zertifikate
Acetate standard solution, 1000 mg/l AcO- in Water (from NaOOCCH₃) standard for ion chromatography
Lieferant: Bernd Kraft
Über diesen Artikel
Frequently Bought Together
Disinfectant solutions, Bacillol AF
Lieferant: BODE CHEMIE
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Lieferant: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Lieferant: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Lieferant: Dr. Weigert
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Lieferant: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Ethanol, GPR RECTAPUR®
Lieferant: VWR Chemicals