Order Entry
Germany
Orders LinkContactUsLinkComponent
Estriol

Estriol

20 µG
Catalog # DIAN12953
Marketsource
Supplier:   DIANOVA
Estriol
Catalog # DIAN12953
Supplier:  DIANOVA
Marketsource
CAS Number:  50-27-1
Estriol

20 µG

CAS Number:  50-27-1
Price
Inquire For Price
Log in for availability
1 * 20 µG
Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • EINECS:200-022-2
Specifications

Specifications

  • Pk:
    20 µG

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.