Order Entry
Germany
ContactUsLinkComponent
E.Z.N.A.® Mollusc & Insect DNA Kit
E.Z.N.A.® Mollusc & Insect DNA Kit
Supplier:  OMEGA BIO-TEK
Certificates
E.Z.N.A.® Mollusc & Insect DNA Kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Invertebrates
  • Target molecule:
    DNA
  • Description:
    Mollusc & Insect DNA Kit

Specifications

Related Information