Burette clamps, Azlon®
Supplier: SciLabware
Certificates
About this item
PP, white. These spring-loaded PP clamps are corrosion resistant and can either hold one or two burettes.
- Allow clear view of graduations along the full length of the burette
- Rubber grips prevent slipping
- Interchangeable compression cone on the central boss provides a firm grip on rods with Ø 11 to 14 mm