Fungal DNA kits, E.Z.N.A.®
Catalog # D3390-02
Supplier: OMEGA BIO-TEK
CAS Number:
Specifications
- Pk:200 T
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:Yeast/Fungi
- Target molecule:DNA
- Description:E.Z.N.A.® Fungal DNA Mini Kit
- Seq:07
- Sample size:100 mg wet or 30 mg dry tissue
- Description:E.Z.N.A.® Fungal DNA Mini Kit
Specifications
About this item
The E.Z.N.A.® Fungal DNA kits allow for the rapid and reliable isolation of high quality total cellular DNA from a wide variety of fungal species without the need for organic extraction.