ContactUsLinkComponent
Cell culture reagents, microcarriers, Cytopore 1
Cell culture reagents, microcarriers, Cytopore 1
Catalog # 17-0911-01
Supplier:  Cytiva
undefined
Cell culture reagents, microcarriers, Cytopore 1
Catalog # 17-0911-01
Supplier:  Cytiva
Specifications
  • Description:
    Cytopore 1, dry powder
  • Pk:
    20 g
Specifications
About this item
Cytopore1 macroporous microcarriers are principally designed for use in suspension culture systems for growth of adherent recombinant Chinese Hamster Ovary (CHO) cells and the production of recombinant proteins.
  • Macroporous structure of Cytopore microcarriers encourages cell growth into the bead, while the micropores provide maximum nutrient availability
  • The characteristics enable growth of cells which require high recirculation rates and high nutrient-availability
  • Optimized for growth of r-CHO in stirred tank cultures but is applicable to cell lines requiring similar surface charge
  • Transparent for easy microscopic examination of attached cells