Orders LinkContactUsLinkComponent
Microarray hybridisation solution v2.0
Microarray hybridisation solution v2.0
Microarray Hybridisation Solution v2.0, 1
Catalog # RPK0325
Supplier:   Cytiva
Microarray hybridisation solution v2.0
Catalog # RPK0325
Supplier:  Cytiva
CAS Number:  
Microarray hybridisation solution v2.0
Microarray hybridisation solution v2.0
Microarray Hybridisation Solution v2.0, 1
CAS Number:  
Price
202,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Microarray hybridisation solution version 2.0, for highly reproducible microarray results.
  • Includes a blend of proprietary components to provide highly reproducible, high-quality automated and manual DNA microarray hybridisations
  • Hybridisation rate enhancers improve the detection of rare targets
  • Proprietary blocking agents minimize non-specific background
DNA microarrays can be used to measure the expression levels of large numbers of genes simultaneously (gene expression profiling), or to genotype multiple areas of a genome (SNP arrays), with variable coverage or resolution depending how many DNA 'spots' cover those areas.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Microarray Hybridisation Solution v2.0
Specifications