Absorption granules - Universal coarse grain
Supplier: asecos
Certificates
About this item
Absorption granules for oil, alkalis, grease, motor fuels, varnishes.
- For outdoor and indoor use
- Officially tested and approved for use on traffic areas
- 20 kg sack binds up to 16 litres of heating oil
- Suitable for solid surfaces
- Even with total saturation of the granules, anti-slipping is guaranteed