Instrument trays
Supplier: VITLAB
Certificates
About this item
Made of melamine, white.
- Tall shape
- Rounded corners
- Smooth surfaces, easy to clean
Practical tray for instruments, tools, and sensitive utensils. Stable and self-supporting.
Ordering information: Fitting lid made of PS, please order separately.