VACUETTE® Urine Collection Tubes
Supplier: Greiner Bio-One
New Product
Certificates
About this item
These sterile tubes are used for urine collection, transportation, centrifugation, strip tests, analysis and storage.
- Exact dosage vacuum and meet quality standards
- With or without preservative
Hermetically sealed, unbreakable and space saving, with improved safety for laboratory and hospital personnel through the use of a closed system, contamination is virtually impossible when decanting into a secondary vessel.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904