Order Entry
Germany
ContactUsLinkComponent
Utility tray with grate, PIG®
Utility tray with grate, PIG®
Catalog # NEWPPAK123-BK
Supplier:  NEW PIG
undefined
Utility tray with grate, PIG®
Catalog # NEWPPAK123-BK
Supplier:  NEW PIG

Specifications

  • Capacity:
    30 l
  • Description:
    Utility tray with grate, LDPE
  • W×D×H:
    640×1250×80 mm
  • Packed:
    1 each
  • Pk:
    1

Specifications

About this item

Durable PE utility tray is ideal for temporary storage of leaking parts or cores awaiting degreasing, repair or pickup. It helps to save bench space as large and heavy parts like batteries, grimy cores waiting for rebuild and leaky containers can be stored safely on the floor.

  • Strong and durable, chemical resistant
  • Ribbed bottom prevents batteries and containers from resting in spilled liquids
  • Heavy-duty walls and ribbed bottom provide strength and prevent warping
  • Includes heavy-duty grate to elevate parts and containers out of deeper liquids in sump

Weight: 11,6 kg