Order Entry
Germany
ContactUsLinkComponent
 
PIG® GRIP-DRI absorbent granules
  NEWPPLPE260
 :  NEW PIG
undefined
PIG® GRIP-DRI absorbent granules
  NEWPPLPE260
 :  NEW PIG

 

  • Description:
    Absorbent granules, Danish moler, non-flammable
  • Absorbency:
    19,8 L
  • Packed:
    1 each
  • Pk:
    1

 

 

Fast acting universal absorbent is chemically inert, making it compatible with almost any liquid.

  • Remains granular, even when saturated, to provide excellent retention; stands up to heavy traffic without releasing liquids
  • Fire-retardant product is ideal for use with low-flash point solvents and other flammables, and in high-temperature applications
  • Micro-granular formulation absorbs 25% more liquid than regular moler absorbents
  • Low-dust and easy to sweep away; won't leave behind an oily sheen or residue, so it helps you create cleaner, safer floors
  • Non-toxic mineral product is safe to handle and use without any risk

Instantly soaks up oils, coolants, solvents, water, acids and caustics.

Weight: 15 kg per bag

Volume bag: 30 L