Order Entry
Germany
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ IgM Purification HP
Affinity chromatography columns, HiTrap™ IgM Purification HP
Catalog # 17-5110-01
Supplier:  Cytiva
undefined
Affinity chromatography columns, HiTrap™ IgM Purification HP
Catalog # 17-5110-01
Supplier:  Cytiva

Specifications

  • Flow rate:
    <4 ml/min
  • Particle Size:
    34 µm
  • Max. pressure:
    5 bar
  • Capacity:
    1 ml
  • Description:
    HiTrap™ IgM Purification HP
  • Seq:
    01
  • pH range:
    3 - 11
  • Storage:
    4 to 8 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7×25 mm
  • Pk:
    5

Specifications

About this item

HiTrap IgM Purification HP is a 1 ml column prepacked with Sepharose High Performance for fast and efficient purification of monoclonal IgM.

  • Optimized for fast, efficient purification of monoclonal IgM from hybridoma cell culture
  • Prepacked with a thiophilic adsorption resin, 2-mercaptopyridine coupled to Sepharose High Performance
  • Recovery of IgM is high with excellent activity retention
  • The top and bottom frits are manufactured from porous polyethylene

Thiophilic adsorption is promoted by water-structuring salts. It has been suggested that the interaction of IgM and ligand results from combined electron donating and accepting action of the ligand or, alternatively, a mixed mode hydrophilic-hydrophobic interaction.

Delivery: Columns are delivered with a stopper on the inlet and a snap-off end on the outlet.