Multimodal chromatography media, Capto™ Core 700
Supplier: Cytiva
Certificates
About this item
Capto™ Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules (molecular weight >700000).
- Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough
- Significantly improved productivity and higher flow rates compared with gel filtration methods
- Straightforward optimisation due to flowthrough chromatography and robust performance
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns
The novel core bead technology and multimodal, octylamine ligand give dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin). Available in various pack sizes for packing columns and in convenient, prepacked formats.
Specifications
- pH range:3 - 13
- Matrix:Highly cross-linked agarose
- Ligand:Octylamine
- Storage temperature:4 - 30 °C
- Binding capacity:~ 13 mg ovalbumin
- Total ionic capacity:40 - 85 µmol/ml (Cl-)
- Flow rate:<500 cm/h