Order Entry
Germany
ContactUsLinkComponent
Ion exchange chromatography media, Capto™ S
Ion exchange chromatography media, Capto™ S
Supplier:  Cytiva
Certificates
undefined
Ion exchange chromatography media, Capto™ S
Supplier:  Cytiva
Certificates

Specifications

  • pH range:
    4 - 12
  • Matrix:
    Highly cross-linked agarose with dextran surface extender
  • Ligand:
    Sulfonate group
  • Storage temperature:
    4 - 30 °C
  • Total ionic capacity:
    0,11 - 0,14 mmol Na+/ml

Specifications