Reaction vessels, cylindrical, with flat flange
DURAN® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With thermostatic and vaccum jacket and withdrawal valve.
Valve bore of 10 mm