Superose™ 6 Gel Filtration Media, Prep Grade
Catalog # 17-0489-01
Supplier: Cytiva
CAS Number:
Specifications
- Particle Size:30 - 40 µm
- Pk:125 ml
- Description:Superose™ 6 (prep grade)
- pH range:3 - 12
- Matrix:Highly cross-linked agarose
- Fractionation range:5 × 103 - 5 × 106 globular proteins
- Storage temperature:4 - 30 °C
- Type:Liquid
Specifications
About this item
Superose™ 6 PG for separations of proteins and other biomolecules according to size. Wide separation in two molecular weight ranges from 1000 up to 5 million.
- Achieve high resolution separations across exceptionally broad molecular weight ranges
- Excellent reproducibility and durability
- High performance gel filtration media specially designed for preparative purification of biomolecules
Low ionic strength eluents (<0,05 M) can utilise a hydrophobic interaction component to alter the selectivity of Superose™ for some lipids, peptides and small aromatic compounds.