Order Entry
Germany
ContactUsLinkComponent
Anion exchange chromatography column, HiScreen™ Capto™ DEAE
Anion exchange chromatography column, HiScreen™ Capto™ DEAE
Catalog # 28-9269-82
Supplier:  Cytiva
undefined
Anion exchange chromatography column, HiScreen™ Capto™ DEAE
Catalog # 28-9269-82
Supplier:  Cytiva

Specifications

  • Ø int.:
    7,70 mm
  • Particle Size:
    90 µm
  • Max. pressure:
    3 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto™ DEAE
  • Length:
    100 mm
  • pH range:
    2 - 9
  • Column hardware:
    PP
  • Pk:
    1

Specifications

About this item

HiScreen™ Capto™ DEAE column is a weak anion exchanger with a bed height of 10 cm making it an excellent tool for method optimisation, parameter screening, robustness testing, and convenient scale-up.

  • Prepacked, ready-to-use columns for convenient process development optimisation
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Easily connected in series to achieve 20 cm bed height
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
  • Used in an optimal way with liquid chromatography systems such as ÄKTA™
  • Flow velocity: <700 cm/h

Process fluid velocities can be applied, since the bed height gives enough residence time and the results can then serve as basis for linear process scale-up. It is prepacked with Capto™ DEAE ion exchange chromatography medium.